HEPES sodium salt ≥98.0%, crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Sodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulphonate, 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, Sodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulphonate, Sodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulfonate, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid) sodium salt, 4-(2-Hydroxyethyl)piperazine-1-ethanesulphonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) sodium salt, HEPES sodium, Sodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulfonate, HEPES Na

0210559325 0210559380 0210559383 0210559391
IC10559325EA 65.05 USD
IC10559325 IC10559380 IC10559383 IC10559391
HEPES sodium salt ≥98.0%, crystalline powder
HEPES sodium salt
Formula: C₈H₁₇N₂NaO₄S
MW: 260.29 g/mol
MDL Number: MFCD00036463
CAS Number: 75277-39-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56632c945c33eb031f86d9a0784e4a3459edfcb8&vid=550cabb1567af1bdb92ab367896c8a9853d7f32d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Synonyms 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt; N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) sodium salt
Form White Powder
Storage Store at Room Temperature (15-30 °C). Store Desiccated.
Loss on Drying 3.0%
pH 9.5 - 10.8(1% aq soln)
pka 7.55 at 25 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR