Tristearin (glycerol tristearate) ~65%, off-white powder, Technical Grade

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Propane-1,2,3-triyl tristearate, Tristearin

0210313105 0210313180
IC10313105EA 269.27 USD
IC10313105 IC10313180
Tristearin (glycerol tristearate) ~65%, off-white powder, Technical Grade
Tristearin (Glycerol tristearate)

Soluble in chloroform (100 mg/mL - clear, almost colorless solution), benzene or hot alcohol; insoluble in water or cold alcohol.


Soluble in chloroform (100 mg/ml - clear, almost colourless solution), benzene or hot alcohol; insoluble in water or cold alcohol.

Formula: C₅₇H₁₁₀O₆
MW: 891.4797 g/mol
Melting Pt: 53…56 °C (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00036230
CAS Number: 555-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR