Sodium borohydride ≥98%, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate

0210289425 0210289450 0210289480 0210289490
IC10289425EA 71.59 USD
IC10289425 IC10289450 IC10289480 IC10289490
Sodium borohydride ≥98%, powder
Sodium borohydride

Soluble in water. Reacts slowly with ethanol. Aqueous solutions are most stable in the presence of small amounts of NaOH (0.2% for a nearly saturated solution containing 44% sodium borohydride) and can be kept for several days. Solutions are rapidly decomposed by boiling.


Sodium borohydride is an excellent reducing agent. It is used to generate diborane, as a foaming agent, as a scavenger for traces of aldehyde, ketones and peroxides in organic chemicals. Nanocrystalline superlattices in gold colloid solution have been prepared by ligand-induction using AuCl3 reduced with sodium borohydride. Nucleophilic addition of hydride ion from sodium borohydride is an inexpensive alternative method for the Baylis-Hillman reaction to form [E]-α-methylcinnamic acids.

Formula: NaBH₄
MW: 37.83 g/mol
MDL Number: MFCD00003518
CAS Number: 16940-66-2
UN: 1426
ADR: 4.3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a366aa43939d54875ac3d0ff56eb96c4824e836&vid=3a366aa43939d54875ac3d0ff56eb96c4824e836&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR