Saponin (from plant), white powder, Practical grade
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Saponin is "built" from quillaic acid, a sugar and possibly other substances. It is a Glycoside widely distributed in plants.
1% of saponin solution can hemolyze erythrocytes. Saponin solutions have been used in the following ways: concentrations of 0.005%-0.01% were used to permeabilize cultured human intestinal epithelial cells after incubation for 30' prior to immunofluorescence; a 0.05% solution of saponin was reportedly used to permeabilize paraformaldehyde-saponin- fixed human fibroblasts for the staining of intracellular fibronectin by different types of peroxidase conjugates for light and electron microscopy; saponins, a surface-active agent, have also been used to lyse the outer membranes of Rous sarcoma viruses, cell membranes of chicken liver and erythrocytes of human and guinea pig. It also has been used as an adjuvant for vaccines.
Potent hemolytic when injected i.v.; surfactant that enhances penetration of proteins and other macromolecules through cell membranes.
Freely soluble in alcohols. Soluble in water.
1% of saponin solution can hemolyze erythrocytes. Saponin solutions have been used in the following ways: concentrations of 0.005%-0.01% were used to permeabilize cultured human intestinal epithelial cells after incubation for 30' prior to immunofluorescence; a 0.05% solution of saponin was reportedly used to permeabilize paraformaldehyde-saponin- fixed human fibroblasts for the staining of intracellular fibronectin by different types of peroxidase conjugates for light and electron microscopy; saponins, a surface-active agent, have also been used to lyse the outer membranes of Rous sarcoma viruses, cell membranes of chicken liver and erythrocytes of human and guinea pig. It also has been used as an adjuvant for vaccines.
Caution: Research Use Only (RUO).
Storage Temperature:
Ambient |
MDL Number:
MFCD00081981 CAS Number: 8047-15-2 |
Specification Test Results
Identity Test | Passes |
Source | Plant |
FTIR | Conforms to Standard |
pH (5% aq soln) | 3-7 |
Loss on Drying | ≤10% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...