L(+)-Histidine monohydrochloride monohydrate, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: L-Histidine monohydrochloride monohydrate, (2S)-(+)-Histidine monohydrochloride monohydrate, L-Histidine hydrochloride monohydrate, (S)-(+)-Histidine monohydrochloride monohydrate, His HCl monohydrate, H-His-OH.HCl.H2O, H-His-OH·HCl·H₂O, L-3-Imidazol-4-ylalanine monohydrochloride monohydrate, L-Histidine hydrochloride hydrate

0210195725 0210195780 0210195790 0210195791
IC10195725EA 36.59 USD
IC10195725 IC10195780 IC10195790 IC10195791
L(+)-Histidine monohydrochloride monohydrate, white powder
L(+)-Histidine monohydrochloride monohydrate

L-Histidine has been used to study cultures of the human T-lymphoblastic leukemia cell line MOLT-4 to study modulation of apoptosis. Exogenous histidine has been shown to enhance the biosynthesis of lovastatin by cultured Aspergillus terreus. Histidine has been utilized as a single nitrogen source to probe swarming in Pseudomonas aeruginosa on agar. An in vivo study has used L-histidine to diminish the net secretory response of the small intestine of of cholera toxin-challenged mice.


Because the pKa of the imidazole group is close to physiological pH, the imidazole moiety can be either uncharged or positively charged, depending on the local environment. This property makes histidine residues of exceptional interest in the active sites of many proteins, such as hemoglobin and myoglobin, where the imidazole ring can readily alternate between the charged (imidazolium) and uncharged (imidazole) states to participate in bond formation and breakage. Histidine is biosynthesized from ATP, 5-phosphoribosyl-1-pyrophosphate (PRPP), and glutamine. In turn, histidine is degraded to glutamate by histidase, urocanase, and imidazolonepropionase via the formation of urocanate, 4-imidazolone 5-propionate, and N-formiminoglutamate.


Soluble in water (50 mg/mL); insoluble in diethyl ether , methanol and alcohol.


L-Histidine has been used to study cultures of the human T-lymphoblastic leukemia cell line MOLT-4 to study modulation of apoptosis. Exogenous histidine has been shown to enhance the biosynthesis of lovastatin by cultured Aspergillus terreus. Histidine has been utilized as a single nitrogen source to probe swarming in Pseudomonas aeruginosa on agar. An in vivo study has used L-histidine to diminish the net secretory response of the small intestine of of cholera toxin-challenged mice.


Caution: Research Use Only (RUO).

Formula: C₆H₉N₃O₂·HCl·H₂O
MW: 209.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151027
CAS Number: 5934-29-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=561d2381f930afe618f78d680790ea9a43f6596c&vid=8aacf6dc4ef5382410cb743498272b48c3c36999&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
pH (2% aq soln) 3 - 5
Loss on Drying (145 - 150°C, 3 hour) 7 - 10%
Optical Rotation +9,5° ± 1° (c = 1, 6 N HCl)
pka 1,80 (COOH); 9,33 (NH2); 6,04 (imidazole group) (Lit.)
Isoelectric Point 7,641(Lit.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR