Sodium-L(+)-glutamate monohydrate ≥99%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: (S)-(+)-2-Aminopentanedioic acid sodium salt monohydrate, Sodium L-Glutamate Monohydrate, L(+)-Glutamic acid sodium salt monohydrate, L(+)-2-Aminopentanedioic acid sodium salt monohydrate, (S)-(+)-Glutamic acid sodium salt monohydrate, L(+)-Glutamic acid monosodium salt monohydrate, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-Sodium glutamate monohydrate, L(+)-α-Glutamic acid sodium salt monohydrate, L(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-α-Glutamic acid sodium salt monohydrate

0210180080 0210180091
IC10180080EA 43.66 USD
IC10180080 IC10180091
Sodium-L(+)-glutamate monohydrate ≥99%, white powder
Sodium-L(+)-glutamate monohydrate

L-Glutamic Acid Monosodium Salt is used as a neurotransmitter at fast synapses; agonist at kainate, NMDA and quisqualate receptors.


Very water soluble; sparingly soluble in alcohol.


L-Glutamic Acid Monosodium Salt is used as a neurotransmitter at fast synapses; agonist at kainate, NMDA and quisqualate receptors.


Caution: Research Use Only (RUO).

Formula: C₅H₈NNaO₄·H₂O
MW: 187.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150138
CAS Number: 6106-04-3
Merck Index: 13,06278

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
Optical Rotation (c=1, 1 N HCl) +20° to +36°
Loss on Drying 0,5%
pH (5% aq soln) 6,7 - 7,2
Solubility (10% aq soln) Clear and Colorless
pka 2,10 (a-COOH), 9,47 (a-NH2), 4,07 (w-COOH) (Lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR