(±)-α-Lipoic acid ≥98%, Powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)-1,2-Dithiolane-3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid
DL-Thioctic Acid is a hydrogen transferrin cofactor.
Antioxidant and coenzyme needed for the activity of enzyme complexes such as those of pyruvate dehydrogenase and glycine decarboxylase. Exogenous thioctic acid is reduced intracellularly by two or more enzymes. The reduced form then influences a number of cell processes by direct radical scavenging, recycling of other antioxidants, increasing glutathione synthesis, and modulating transcription factor activity particularly that of NF-κB. Decreases the phagocytosis of myelin by macrophages.
Soluble in fat solvents, methanol (50 mg/mL-clear pale yellow solution), ethanol(50 mg/mL-clear solution); practically insoluble in water.
DL-Thioctic Acid is a hydrogen transferrin cofactor.
Formula:
C₈H₁₄O₂S₂ MW: 206.33 g/mol |
MDL Number:
MFCD00005474 CAS Number: 1077-28-7 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...