Cysteaminium chloride ∼98%, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
Cysteamine Hydrochloride
, Mercaptoethylamine-HCl
, β-Mercaptoethylamine hydrochloride
, H-Cys-NH2·HCl
, 2-Aminoethanethiol hydrochloride
, 2-MEA
, 2-Mercaptoethylamine hydrochloride
β-Mercaptoethylamine (Cysteamine hydrochloride) is useful antioxidant. It is experimentally used as a radioprotective agent. It is also used to produce acute and chronic duodenal ulcers in rats and as an antidote to acetaminophen.
Soluble in water (50 mg/mL) clear,colorless to faint yellow,Soluble in alcohol.
Caution: Research Use Only (RUO).
Formula:
C₂H₇NS·HCl MW: 113.61 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00012904 CAS Number: 156-57-0 |
Specification Test Results
Identity Test | Passes |
Purity | ~98% |
FTIR | Conforms to Standard |
Melting Point | 58 - 73 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...