Anthrone, dark yellow powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
Anthracen-9(10H)-one
0210069110
0210069125
IC10069110EA
56.31
USD
IC10069110
IC10069125
Anthrone, dark yellow powder
Anthrone
Anthrone is a tricyclic aromatic hydrocarbon.
It is used to determine hydrocarbons using a colorimetric method. It helps to estimate carbohydrates in yeast and plant extracts.
Soluble in ethyl acetate.
Anthrone is a tricyclic aromatic hydrocarbon.
Caution: Protect from air oxidation
Formula:
C₁₄H₁₀O MW: 194.23 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00001187 CAS Number: 90-44-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...