Caprylic acid ≥99.5%

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Octanoic acid, n-Octanoic Acid, Octoic acid

0210039125 0210039190 0210039191 0210039180
IC10039125EA 84.06 USD
IC10039125 IC10039190 IC10039191 IC0210039180
Caprylic acid ≥99.5%
Caprylic acid

Caprylic acid is the common name for the eight-carbon saturated fatty acid known by the systematic name octanoic acid. It is found naturally in the milk of various mammals, and it is a minor constituent of coconut oil and palm kernel oil.
Caprylic acid is an intermediate in the manufacture of perfume esters and of dyes.


Caprylic acid is the common name for the eight-carbon saturated fatty acid known by the systematic name octanoic acid. It is found naturally in the milk of various mammals, and it is a minor constituent of coconut oil and palm kernel oil.

Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 239.7 (lit.)
Melting Pt: 16 (lit.)
Density: 0.91 g/cm³ (20 °C) (lit.)
Flash Pt: 130
Storage Temperature: Ambient
MDL Number: MFCD00004429
CAS Number: 124-07-2
UN: 3265
ADR: 8 III
Merck Index: 13,01771
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c3030695a3c15cc43fd4dc3c61d5b3356b4291e&vid=2c3030695a3c15cc43fd4dc3c61d5b3356b4291e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity 99.6%
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR