Adonitol, white powder

Supplier: 0000012207

0210025205 0210025225 0210025280
IC10025205EA 42.94 USD
IC10025205 IC10025225 IC10025280
Adonitol, white powder
Adonitol

Adonitol (ribitol), a pentose alcohol, is metabolised to teicholic acids used in the cell walls of gram positive bacteria. Adonitol is often compared to other cell permeating molecules such as formamide, propanediol, and DMSO as a cryopreservation agent.


Adonitol is a crystalline pentose alcohol (C5H12O5) formed by the reduction of ribose which does not reduce benedict solution.


Soluble in water (100 mg/ml - clear, colorless solution)


Adonitol is a crystalline pentose alcohol (C5H12O5) formed by the reduction of ribose which does not reduce benedict solution.


Caution: Research Use Only (RUO).

Formula: HOH₂C(CH(OH))₃CH₂OH
MDL Number: MFCD00064291
CAS Number: 488-81-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8d1883835f868a432133e4feeed1fff21350ed5&vid=f8d1883835f868a432133e4feeed1fff21350ed5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Melting Point 98 - 106 °C
FTIR Conforms to Standard
Moisture Content 0.50%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR