5-Bromo-2′-deoxycytidine hydrate, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210016680 0210016691
IC10016680EA 57.6 USD
IC10016680 IC10016691
5-Bromo-2′-deoxycytidine hydrate, white powder
5-Bromo-2′-deoxycytidine hydrate

5-Bromo-2'-deoxycytidine can be incorporated into DNA in the presence of DNA polymerase, replacing dCTP.


Soluble in water, NH₄OH (50 mg /ml).

Formula: C₉H₁₂BrN₃O₄·×H₂O
MW: 306.116 g/mol (anhy)
Storage Temperature: Freezer
CAS Number: 1022-79-3

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
UV/VIS: λmax (water) 286 - 289 nm
Moisture Report Result


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR