Kanamycin sulfate 50 mg/mL in distilled water, sterile filtered

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Kanamycin Monosulfate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulfate, Kanamycin A, Kanamycin Monosulphate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphate

J67354-AD J67354-AE
AAJ67354-ADEA 298.41 USD
AAJ67354-AD AAJ67354-AE
Kanamycin sulfate 50 mg/mL in distilled water, sterile filtered
Kanamycin sulphate

Kanamycin, 50 mg/mL in distilled water, sterile-filtered.


  • Liquid


Kanamycin is used in tissue culture media and as an additive in culture media for the isolation of group D Streptococci on Kanamycin Esculin Azide Agar and for selection of transformed plant cells containing the neomycin phosphotransferase on a kanamycin-medium.


This antibiotic can also be used as a selection agent for cells transformed with kanamycin B resistance gene and in several types of isolation media. It is recommended for use in cell culture applications at 100 mg/mL.


Fully miscible in water, insoluble in acetone, chloroform, ether, ethyl acetate, and alcohol


Caution: Research use only: Not intended for animal or human diagnostic or therapeutic use.

Formula: C₁₈H₃₈N₄O₁₅S
MW: 582.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00070253
CAS Number: 25389-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR