Polymyxin B sulphate 50 mg/mL in distilled water, sterile filtered

Supplier: Thermo Scientific Chemicals

J67163-8EQ J67163-XF
AAJ67163-8EQEA 307 USD
AAJ67163-8EQ AAJ67163-XF
Polymyxin B sulphate 50 mg/mL in distilled water, sterile filtered
Polymyxin B sulphate
Formula: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
MW: 1385.61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00131991
CAS Number: 1405-20-5
EINECS: 215-774-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR