Pimaricin 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Natamycin
Natamycin (Pimaricin) is an amphoteric antibiotic from Streptomyces chattanoogensis
- Light sensitive
Off-white powder
It is used as an ergosterol and cholesterol binding agent to study lipid bilayer dynamics. It is used to study the pleitropic regulator AdpAch and the gamma-butyrolactone regulatory system of Streptomyces chattanoogensis.
Soluble in DMSO
Caution: Research use only: Not intended for animal or human diagnostic or therapeutic use.
Formula:
C₃₃H₄₇NO₁₃ MW: 665.733 g/mol Boiling Pt: 952 °C Melting Pt: 280 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00135085 CAS Number: 7681-93-8 EINECS: 231-683-5 Merck Index: 14,6427 |
Specification Test Results
Assay | 99% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...