N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Fmoc-L-aspartic Acid, Fmoc-Asp-OH
H62274-06
H62274-14
AAH62274-06EA
42.2
USD
AAH62274-06
AAH62274-14
AAH62274-22
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 95%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid
N-Fmoc-L-aspartic acid, 95%
Purity: 95%
CAS No.: 119062-05-4
Molecular Formula: C19H17NO6
Formula Weight: 355.34
Storage Temperature: -30°C to -10°C
Beilstein Reference: 3632557
MDL No.: MFCD00237654
Formula:
C₁₉H₁₇NO₆ MW: 355.35 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00237654 CAS Number: 119062-05-4 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...