Gaumard® Newborn Patient Care Manikin
Supplier: American 3B Scientific
1005802 1005805
470119-304EA 1094.99 USD
470119-304 470119-306
Gaumard® Newborn Patient Care Manikin
Models Human Biology Models
Advanced Infant Nursing Care Manikin
  • Heel stick and finger prick technique
  • Soft upper body skin over torso for “baby like” feel
  • Bathing and bandaging activity
  • Intramuscular injection in upper thigh
  • Interchangeable genitalia
  • Male and female catheterization
  • Enema administration
This manikin simulates a 0-8 week old newborn with rotatable soft arms and legs. It allows training of basic and advanced pediatric patient care procedures such as: Bathing and bandaging, Tube feeding and gastric suction (movable tongue), Heel stick and finger prick practice, Male and female catheterization, Enema administration. Supplied with T-Shirt, diaper and carrying bag.