Petri Dishes, Advantec
Supplier: Advantec MFS
42004A02 42004A00 42004A01 42004A03
101105-260PK 222.48 USD
101105-260 101105-220 101105-292 101105-740
Petri Dishes, Advantec
Culture Dishes Petri Dishes
Polystyrene dishes suitable for culturing microorganisms on 47 mm diameter membrane filters
Dishes are sterilized by gamma irradiation (50 x 11) or by EtO. Standard size 50 x 11 mm polystyrene dishes fit all commercially available aluminum block incubators. Dishes are available with or without 47 mm absorbent cellulose pad (0.85 ± 0.17 mm thick, absorbs 1.8-2.2 mL liquid).