Anti-UBASH3A Rabbit Polyclonal Antibody

Supplier: Proteintech
15823-1-AP-150UL
10095-414EA 453 USD
10095-414
Anti-UBASH3A Rabbit Polyclonal Antibody
Antibodies
UBASH3A gene encodes ubiquitin-associated and SH3 domain-containing protein A (UBASH3A) also known as T-cell ubiquitin ligand 1 (TULA1) which is one of two family members belonging to the T-cell ubiquitin ligand (TULA) family. UBASH3A gene expression is limited to only a few tissues, with its highest expression in spleen, peripheral blood leukocytes, and bone marrow. UBASH3A protein is associated with c-Cbl and ubiquitylated proteins and has been implicated in the regulation of signaling mediated by protein-tyrosine kinases oth family members can negatively regulate T-cell signaling. UBASH3A can facilitate growth factor withdrawal-induced apoptosis in T cells via its interaction with apoptosis-inducing factor (AIF). This antibody is UBASH3A-specific.

Western Blot: Human spleen Tissue, 1:200-1:2000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: UBASH3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR