Sodium thiosulfate, anhydrous ≥97.0% (by iodometry), Supelco®

Supplier: MilliporeSigma

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite

M-Clarity™ quality level = MQ200

1.06512.0250 1.06512.2500 1.06512.9025 1.06512.9050
EM1.06512.0250CS 3819 USD
EM1.06512.0250 EM1.06512.2500 EM1.06512.9025 EM1.06512.9050
Sodium thiosulfate, anhydrous ≥97.0% (by iodometry), Supelco®
Sodium thiosulphate
Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100
Melting Pt: 48
Density: 1.67 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR