Cyclohexane ≥99% (by GC), EMPLURA®, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Hexahydro-Benzene, Hexahydrobenzene, Hexamethylene
M-Clarity™ quality level = MQ200
1.02832.1000
1.02832.2500
1.02832.6025
1.02832.6190
1.02832.9190
EM1.02832.1000CS
511
USD
EM1.02832.1000
EM1.02832.2500
EM1.02832.6025
EM1.02832.6190
EM1.02832.9190
Cyclohexane ≥99% (by GC), EMPLURA®, Supelco®
Cyclohexane
Formula:
C₆H₁₂ MW: 84.16 g/mol Boiling Pt: 80.7 °C (lit.) Melting Pt: 4…7 °C (lit.) Density: 0.779 g/ml (25°C) (lit.) Flash Pt: –18 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003814 CAS Number: 110-82-7 EINECS: 203-806-2 UN: 1145 ADR: 3,II Merck Index: 13,02752 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...