Tetrahydrofuran, LiChrosolv® for liquid chromatography, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Oxolane, 1,4-epoxy-Butane, Butylene oxide, Butane alpha,delta-oxide, Tetramethylene oxide, THF, Oxacyclopentane, Cyclotetramethylene oxide, Diethylene oxide

M-Clarity™ quality level = MQ100

1.08101.1000 1.08101.2500 1.08101.4000
EM1.08101.1000CS 1438 USD
EM1.08101.1000 EM1.08101.2500 EM1.08101.4000
Tetrahydrofuran, LiChrosolv® for liquid chromatography, Supelco®
Tetrahydrofuran

Offering a high degree of UV transmittance, low particle count, low acidity and alkalinity, and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. They are produced from specially selected raw materials, and undergo a number of purification steps prior to final packaging. Since separations are normally carried out under gradient conditions in analytical HPLC, Merck Millipore offer solvents in ’gradient grade’ as well as ’isocratic grade’. This enables you to minimise the gradient effect of the solvent involved.


Offering a high degree of UV transmittance, low particle count, low acidity and alkalinity, and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. They are produced from specially selected raw materials, and undergo a number of purification steps prior to final packaging. Since separations are normally carried out under gradient conditions in analytical HPLC, Merck Millipore offer solvents in ’gradient grade’ as well as ’isocratic grade’. This enables you to minimise the gradient effect of the solvent involved.


The LC-MS LiChrosolv® hypergrade quality solvents are accurately tested for LC-MS suitability, and meet all the requirements of modern LC-MS ionisation methods (ESI/ APCI – positive and negative mode) with low level of ionic background and low ion suppression, they ensure high reproducibility and high ionisation efficiency.

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: –108.5 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -108.5 °C
Molar Mass 72.11 g/mol
Boiling Point 65 - 66 °C (1013 hPa)
Vapor Pressure 173 hPa (20 °C)
Flash Point -20 °C
Refractive Index
Explosion Limit 1.5 - 12.4 %(V)
Density 0.89 g/cm3 (20 °C)
pH 7 - 8 (200 g/l, H2O, 20 °C)
Ignition Point 215 °C
Viscosity kinematic
Synonyms THF, Tetramethylene oxide, Oxolane, Butylene oxide
Hill Formula C4H8O
Chemical Formula

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR