Ethanol ≥99.9% (by GC), LiChrosolv®, gradient grade for liquid chromatography, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Ethyl alcohol

M-Clarity™ quality level = MQ100

1.11727.1000 1.11727.2500 1.11727.4000 1.11727.0500
EM1.11727.1000CS 2513 USD
EM1.11727.1000 EM1.11727.2500 EM1.11727.4000 EM1.11727.0500
Ethanol ≥99.9% (by GC), LiChrosolv®, gradient grade for liquid chromatography, Supelco®
Ethanol

Offering a high degree of UV transmittance, low particle count, low acidity and alkalinity, and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. They are produced from specially selected raw materials, and undergo a number of purification steps prior to final packaging. Since separations are normally carried out under gradient conditions in analytical HPLC, Merck Millipore offer solvents in ’gradient grade’ as well as ’isocratic grade’. This enables you to minimise the gradient effect of the solvent involved.


The LC-MS LiChrosolv® hypergrade quality solvents are accurately tested for LC-MS suitability, and meet all the requirements of modern LC-MS ionisation methods (ESI/ APCI – positive and negative mode) with low level of ionic background and low ion suppression, they ensure high reproducibility and high ionisation efficiency.

Formula: C₂H₅OH
MW: 46.07 g/mol
Boiling Pt: 78 °C (1013 hPa)
Melting Pt: –114.5 °C
Density: 0.79 g/cm³ (20 °C)
Flash Pt: 12 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,

Order Now

Specification Test Results

Purity (GC) ≥ 99.9 %
Identity (IR) conforms
Evaporation residue ≤ 2.0 mg/l
Water ≤ 0.1 %
Acidity ≤ 0.0002 meq/g
Alkalinity ≤ 0.0002 meq/g
Gradient grade (at 235 nm) ≤ 5 mAU
Gradient grade (at 254 nm) ≤ 2 mAU
Transmission (at 225 nm) ≥ 60 %
Transmission (at 240 nm) ≥ 85 %
Transmission (from 260 nm) ≥ 98 %
Filtered by 0.2 µm filter

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR