Whatman™ Grade 93 Qualitative Filter Papers
Supplier: Cytiva
1093-111 1093-126 1093-125 1093-6215 1093-930 1093-935 1093-110
28449-026PK 224.82 USD
28449-026 28449-048 10035-242 10035-246 10035-248 10035-250 10035-714
Whatman™ Grade 93 Qualitative Filter Papers
Filters Filter Paper Qualitative Filter Paper
These cellulose filter papers are intermediate in speed and retention between grades 1 and 4.
These extremely strong filter papers have a high wet strength due to the addition of a small quantity of chemically stable resin. Normal qualitative applications will not introduce any significant impurities into the filtrate. The resins do, however, contain nitrogen so these grades should not be used in Kjeldahl estimations, etc.
Available in a dispenser pack, which can be attached to the wall or bench, placed on a shelf either upright or flat, and used as a normal carton or as a convenientMore Product Information