Micro-Touch® Nitrile Disposable Examination Gloves
Supplier: Ansell Healthcare
6034300 6034301 6034302 6034304 6034303
89176-406CS 362.22 USD
89176-406 89176-408 89176-410 89176-414 89176-412
Micro-Touch® Nitrile Disposable Examination Gloves
Gloves
Gloves offering exceptional fit and reliable protection, designed for use in healthcare settings.
  • Comfortable fit and feel, providing lightweight protection
  • Ideal for all clinical settings
  • Tested against chemotherapy drugs
  • Excellent barrier integrity (1.5 AQL)
  • Smart packaging; 200-count packaging reduces waste
Lightweight, high-quality nitrile formulation ensures a comfortable fit and feel for healthcare professionals, offering comfortable protection throughout extended use. The gloves provide excellent barrier integrity, with an AQL better than industry standards (1.5) to ensure fewer pinholes defects. They are tested and approved for use with chemotherapy drugs.
Certifications: ASTM D6319, Medical Device Regulation (EU) 2017/745 Class I, FDA21 CFR 177-2600-US Food Contact Approved, ISO 10993-10:2002, ISO 13485, ISO 9001, Food Type: Rubber.
Packaging: Gloves are packaged 200 pieces per dispenser, 10 dispensers per case.