Zinc chloride, anhydrous, Reagent Grade

Supplier: VWR
Danger

Synonyms: Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride

N882-100G N882-500G
89230-044CS 712.77 USD
89230-044 89230-046
Zinc chloride, anhydrous, Reagent Grade
Zinc chloride
Formula: ZnCl₂
Boiling Pt: 732 °C (1013 hPa)
Melting Pt: 283 °C
Density: 2.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 13,10185

Order Now

Specification Test Results

Ammonium (NH4) 0.5%
Nitrate (NO3) 0.003 %
Sulfate (SO4) 0.05 %
Zinc Chloride (ZnCl2) 98 %
Zinc Oxide (ZnO) 0.6 % - 0.7 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR