Trifluoroacetic acid for biotechnology
Supplier: VWR
|
Danger
|
Synonyms:
TFA
0993-100ML
0993-500ML
89399-844CS
1933.32
USD
89399-844
89500-554
Trifluoroacetic acid for biotechnology
Trifluoroacetic acid
Formula:
C₂HF₃O₂ MW: 114.02 g/mol Boiling Pt: 71.8 °C (1013 hPa) Melting Pt: –15 °C Density: 1.490 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004169 CAS Number: 76-05-1 EINECS: 200-929-3 UN: 2699 ADR: 8,I Merck Index: 13,09754 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...