STAR™ DISH Petri Dishes, Single Membrane Filters, Phoenix
Supplier: Phoenix Biomedical
058 057
25389-004CS 308.42 USD
25389-004 25389-006
STAR™ DISH Petri Dishes, Single Membrane Filters, Phoenix
Culture Dishes Petri Dishes
These dishes save space in refrigerators and incubators
They are ideal when small quantities of media are used. They accommodate a single membrane filter for water and pharmaceutical testing applications.