Human Recombinant TPO (from E. coli)
Supplier: Prosci
40-585 40-610
10072-200EA 305 USD
10072-200 10073-102
Human Recombinant TPO (from E. coli)
Proteins and Peptides
TPO is a lineage specific growth factor, produced in the liver, kidney and skeletal muscle. It stimulates the proliferation and maturation of megakaryocytes, and promotes increased circulating levels of platelets in vivo. TPO signals through the c-mpl receptor and acts as an important regulator of circulating platelets. Human and murine TPO exhibits cross-species reactivity. Recombinant human TPO is a fully biologically active 174 amino acid polypeptide (18.6 kDa), which contains the erythropoietin-like domain of the full length TPO protein. Recombinant rat TPO is a fully biologically active 174 amino acid polypeptide (18.7 kDa), which contains the erythropoietin-like domain of the full length TPO protein.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.