Anti-IL1F5 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
201-401-B77 201-401-B77S
RL201-401-B77EA 551.64 USD
RL201-401-B77 RL201-401-B77S
Anti-IL1F5 Rabbit Polyclonal Antibody
Antibodies
This Protein-A purified antibody is suitable for ELISA and Western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 17.1 kDa in size corresponding to bovine IL-1F5 Protein by western blotting.

Recommended Dilutions: ELISA: 8000; Western Blot: 1 µg/ml;

Type: Primary
Antigen: IL1F5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR