Anti-GOX Sheep Polyclonal Antibody

Supplier: Rockland Immunochemical
200-6137-0100 200-6137S
RL20061370100EA 551.64 USD
RL20061370100 RL2006137S
Anti-GOX Sheep Polyclonal Antibody
Antibodies
GLUCOSE OXIDASE antibody has been assayed against 1.0 ug of Glucose Oxidase [Aspergillus niger] in a standard sandwich ELISA using Peroxidase conjugated Affinity Purified anti-Sheep IgG [H&L] (Rabbit) and (ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: User Optimized

Type: Primary
Antigen: gox
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Aspergillus niger
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR