Anti-Avidin Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-4197-0100 200-4197S
RL20041970100EA 380.3 USD
RL20041970100 RL2004197S
Anti-Avidin Rabbit Polyclonal Antibody
Antibodies
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

  • Recommended Dilutions: ELISA: 1:2000 - 1:20000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:1000 - 1:5000

Type: Tag
Antigen: Avidin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR