Anti-UREA_CANEN Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-4182-0100 200-4182S
RL20041820100EA 551.64 USD
RL20041820100 RL2004182S
Anti-UREA_CANEN Rabbit Polyclonal Antibody
Antibodies
Anti-UREASE Antibody has been assayed against 1.0 ug of Urease [Jack Bean] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG [H&L] (Goat) and (ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100

Type: Primary
Antigen: Urease
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Jack Bean
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR