Anti-CELA1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-4168-0100 200-4168S
RL20041680100EA 551.64 USD
RL20041680100 RL2004168S
Anti-CELA1 Rabbit Polyclonal Antibody
Antibodies
ELASTASE atibody has been assayed against 1.0 ug of Elastase [Porcine Pancreas] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG [H&L] (Goat) and (ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:500 - 1:3000; Immunofluorescence: 1:100; Western Blot: 1:500 - 1:3000

Type: Primary
Antigen: CELA1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR