Anti-ARG1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-4142-0100 200-4142S
RL20041420100EA 551.64 USD
RL20041420100 RL2004142S
Anti-ARG1 Rabbit Polyclonal Antibody
Antibodies
ARGINASE Antibody has been assayed against 1.0 µg of Arginase [Bovine Liver] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG [H&L] (Goat) and (ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000

Type: Primary
Antigen: ARG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR