Anti-VEGFA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-401-B74 200-401-B74S
RL200-401-B74EA 551.64 USD
RL200-401-B74 RL200-401-B74S
Anti-VEGFA Rabbit Polyclonal Antibody
Antibodies
This Protein-A purified antibody has been tested for use in ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 19.2 kDa in size corresponding to bovine VEGF-A Protein by western blotting.

Recommended Dilutions: Western Blot: 1:333;

Type: Primary
Antigen: VEGFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR