Anti-GDF15 Mouse Monoclonal Antibody [clone: 23B3D2.H5]

Supplier: Rockland Immunochemical
200-301-B07 200-301-B07S
RL200-301-B07EA 565.22 USD
RL200-301-B07 RL200-301-B07S
Anti-GDF15 Mouse Monoclonal Antibody [clone: 23B3D2.H5]
Antibodies
This Protein A purified antibody is suitable for ELISA and western blotting of human NAG-1 Protein.  For detection of NAG-1 in human serum, a sandwich ELISA is suggested using this antibody in combination with anti-NAG-1/GDF15 (N-terminal), H variant or D.

Recommended Dilutions: ELISA: 1:2000; Western Blot: 1:1000;

Type: Primary
Antigen: NAG-1
Clonality: Monoclonal
Clone: 23B3D2.H5
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR