Anti-PFKM Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
200-1156-0100 200-1156S
RL20011560100EA 551.64 USD
RL20011560100 RL2001156S
Anti-PFKM Goat Polyclonal Antibody
Antibodies
FRUCTOSE-6-PHOSPHATE KINASE antibody has been tested for use in ELISA, immunofluorescence microscopy and western blot.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 48 kDa in size.

Recommended Dilutions: ELISA: 1:10000 - 1:40000; Immunofluorescence: 1:200 - 1:2000; Western Blot: 1:500 - 1:2000; Other Dilution: User Optimized

Type: Primary
Antigen: PFKM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR