Anti-CA1 Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
200-1154-0100 200-1154S
RL20011540100EA 551.64 USD
RL20011540100 RL2001154S
Anti-CA1 Goat Polyclonal Antibody
Antibodies
This antibody is useful in immunohistochemistry. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:2000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunochemistry: 10µg/ml; Other Dilution: User Optimized

Type: Primary
Antigen: CA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR