GeneMate 81-Place Freezer Rack with Lid, Scientific Specialties
Supplier: GeneMate
490004-154 490004-156 490004-158 490004-160 490004-162 490004-164 490004-166 490004-168
490004-154BX 152.96 USD
490004-154 490004-156 490004-158 490004-160 490004-162 490004-164 490004-166 490004-168
GeneMate 81-Place Freezer Rack with Lid, Scientific Specialties
Racks Cryogenic Vial Racks
The 5.25x5.25" square rack, rated to –90°C (–130°F), fits into existing standard freezer storage modules and will accommodate all 1.5/2mL microcentrifuge tubes and standard cryo vials. The grid is alphanumerically matrixed with the 1–9 and A–I characters hot stamped directly onto the grids in a distinctive contrasting color for fast, easy, and error-free identification of individual tube samples.
  • Ideal for 1.5/2 mL microtubes and 1.2 mL cryo vials
  • Sturdy, reusable, polypropylene 81-place cryogenic storage rack
  • Colorful economic alternative to the "disposable" cardboard freezer box
  • Lids also make good staining trays
  • Clear lid with frosted writing panels on two sides