GeneMate Blue Autoradiography Film
Supplier: GeneMate
490001-926 490001-928 490001-930 490001-934 490001-946 490001-950 490001-952 490014-238
490001-926BX 344.26 USD
490001-926 490001-928 490001-930 490001-934 490001-946 490001-950 490001-952 490014-238
GeneMate Blue Autoradiography Film
Autoradiography Films Films
This is outstanding film for the laboratory that covers diverse applications including chemiluminescence and autoradiography of 32P and 125I
Exceptional sharpness and resolution and extremely low backgrounds lead to publishing-grade results. Manual or automatic development consistently superior results for exposure of blotting experiments. Sharp and clear results, box after box.
Three double emulsion blue autorad films available including Blue Basic for ideal for autoradiography and chemiluminescence, Blue Ultra for the highest level of sensitivity for all application and Blue Lite,More Product Information