Supermax Amazing™ Nitrile Gloves
Supplier: SUPERMAX HEALTHCARE INC
92885 92886 92887 92888 92889
75997-412CS 644.57 USD
75997-412 75997-414 75997-416 75997-418 75997-420
Supermax Amazing™ Nitrile Gloves
Gloves
Disposable, non sterile nitrile gloves.
  • 2 mil palm thickness
  • Powder-free
  • Ambidextrous
  • Textured fingers, beaded cuff
Packaging: 3000 gloves per case.