Anti-VWF Mouse Monoclonal Antibody [clone: F8/86]

Supplier: Diagnostic BioSystems

MOB196 PDM019 MOB196-05
95024-550EA 298.1 USD
95024-550 99990-410 99990-850
Anti-VWF Mouse Monoclonal Antibody [clone: F8/86]
Antibodies
This antibody reacts with Von Willebrand factor in endothelial cells. It also reacts with megakaryocytes in human bone marrow.

Type:
Antigen: VWF
Clonality: Monoclonal
Clone: F8/86
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR