Anti-Tyrosinase (Melanoma Marker) Mouse Monoclonal Antibody [clone: T311]

Supplier: Diagnostic Biosystems

MOB290-05 MOB290
95024-872EA 552.33 USD
95024-872 99990-548
Anti-Tyrosinase (Melanoma Marker) Mouse Monoclonal Antibody [clone: T311]
Antibodies
This antibody is specific to a cluster of proteins between 70-80 kDa known as tyrosinase. This antibody does not show any cross-reaction with MAGE-1 and tyrosinase-related protein-1, TRP-1/gp75. Staining of melanomas with this antibody showed tyrosinase in melanotic as well as amelanotic variants.

Type:
Antigen: Tyrosinase (Melanoma Marker)
Clonality: Monoclonal
Clone: T311
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR