Anti-Mast Cell Tryptase Mouse Monoclonal Antibody [clone: AA1]

Supplier: Diagnostic Biosystems

MOB347-05 MOB347 PDM160
95024-660EA 505.96 USD
95024-660 99990-650 10065-082
Anti-Mast Cell Tryptase Mouse Monoclonal Antibody [clone: AA1]
Antibodies
This antibody reacts with mast cells distributed in skin, synovium, lung, and heart. This antibody does not bind with any other cell type. Mast cells contain a number of preformed chemicals mediators such as histamine, chymase, carboxypeptidase, and proteolytic tryptase. Human mast cell tryptase is considered to be an important marker of mast cell activation and is an important mediator of inflammation.

Type:
Antigen: Mast Cell Tryptase
Clonality: Monoclonal
Clone: AA1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR