Anti-KI67 Rabbit Monoclonal Antibody [clone: SP6]

Supplier: Diagnostic Biosystems

RMAB004 RMPD004
99991-134EA 1158.58 USD
99991-134 99991-170
Anti-KI67 Rabbit Monoclonal Antibody [clone: SP6]
Antibodies
Ki-67 is a nuclear protein, which is expressed in proliferating cells. Ki-67 is preferentially expressed during late G1-, S-, M-, and G2-phases of the cell cycle, while cells in the G0 (quiescent) phase are negative for this protein.

Type:
Antigen: KI67
Clonality: Monoclonal
Clone: SP6
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR