Anti-Inhibitin-alpha Mouse Monoclonal Antibody [clone: R1]

Supplier: Diagnostic Biosystems

MOB435 MOB435-05 PDM178
99990-802EA 863.61 USD
99990-802 95024-604 10065-000
Anti-Inhibitin-alpha Mouse Monoclonal Antibody [clone: R1]
Antibodies
Inhibin is a dimeric glycoprotein hormone from the TGF-β family made of α and β subunits. This antibody recognizes the 32kDa α subunit of human inhibin. Inhibin α is expressed in a wide variety of human tissues, such as brain, prostate, adrenal, granulose cells of ovary, sertoli cells of testis and in fetoplacental unit.

Type:
Antigen: Inhibitin-alpha
Clonality: Monoclonal
Clone: R1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR