Anti-IgA Polyclonal Antibody

Supplier: Diagnostic Biosystems

RP020-05 PDR017 RP020
95025-054EA 212.44 USD
95025-054 99991-074 99991-236
Anti-IgA Polyclonal Antibody
Antibodies
This antibody reacts with α-chains of human IgA. This antibody has been adsorbed to a very high level of specificity.

Type:
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR