Anti-Papiloma Virus Type 16 Mouse Monoclonal Antibody [clone: CAMVIR-1]

Supplier: Diagnostic Biosystems

MOB394-05 MOB394 PDM166
95024-592EA 352.32 USD
95024-592 99990-730 10064-984
Anti-Papiloma Virus Type 16 Mouse Monoclonal Antibody [clone: CAMVIR-1]
Antibodies
This antibody stains with HPV type 16. L1 protein is a major capsid protein of HPV type 16. HPV 6 and 11 have been associated with relatively benign diseases such as genital warts but type 16 and 18 are strongly associated with cervical, vaginal and vulvar malignancies.

Type:
Antigen: Papiloma Virus Type 16
Clonality: Monoclonal
Clone: CAMVIR-1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR