Anti-Heliocobacter Pylori Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP016-05 RP016 PDR053
95025-042EA 279.49 USD
95025-042 99991-228 10065-012
Anti-Heliocobacter Pylori Polyclonal Antibody
Antibodies
This antibody reacts with the whole H. pylori. The antibody stains the bacteria present on the surface of the epithelium or in the cytoplasm of the epithelial cells. H. pylori is known to cause peptic ulcer and chronic gastritis in humans.
Type:
Antigen:
Clonality:
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now